missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Sulfosalicylic Acid Dihydrate, 98%
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Supplier: Thermo Scientific Chemicals 132862500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| 5-Sulfosalicylic acid dihydrate | |
| 97.5 | |
| 106.0°C to 110.0°C | |
| Authentic | |
| Plastic bottle | |
| MFCD00007508,MFCD00149540 | |
| 11, 411 | |
| 15, 9094 | |
| Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether | |
| C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O | |
| 250.27 | |
| 254.22 | |
| Crystalline Powder or Crystals |
| 5965-83-3,97-05-2 | |
| 100.0 | |
| White | |
| 98% | |
| C9H14O6S | |
| 250 g | |
| 01,1118 | |
| 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd | |
| NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
| 2-hydroxy-5-sulfobenzoic acid; bis(methane) | |
| 2723734 | |
| 98% |
RUO – Research Use Only