missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic Acid Dihydrate, 98%

Catalog No. p-3737430
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132862500 250 g Plastic bottle
AC132860010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC132862500 Supplier Thermo Scientific Chemicals Supplier No. 132862500
Only null left

CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Specifications

Chemical Name or Material 5-Sulfosalicylic acid dihydrate
CAS 5965-83-3, 97-05-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 106.0°C to 110.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C9H14O6S
Quantity 250 g
MDL Number MFCD00007508,MFCD00149540
Beilstein 11, 411
Fieser 01,1118
Merck Index 15, 9094
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
Solubility Information Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
Molecular Weight (g/mol) 250.27
PubChem CID 2723734
Formula Weight 254.22
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Show More Show Less

RUO – Research Use Only