missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione 93.0+%, TCI America™
Supplier: TCI America P16351G
Specifications
| 5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione | |
| 51°C | |
| 1 g | |
| CCCC1SC2=C(S1)SC(=S)S2 | |
| 252.441 | |
| 252.44 | |
| Crystalline Powder |
| 202126-48-5 | |
| C7H8S5 | |
| QVGTUNLOIXSBRJ-UHFFFAOYSA-N | |
| 2-propyl-[1,3]dithiolo[4,5-d][1,3]dithiole-5-thione | |
| 44629778 | |
| ≥93.0% (HPLC) |
Chemical Identifiers
| 202126-48-5 | |
| 252.441 | |
| 44629778 | |
| CCCC1SC2=C(S1)SC(=S)S2 |
| C7H8S5 | |
| QVGTUNLOIXSBRJ-UHFFFAOYSA-N | |
| 2-propyl-[1,3]dithiolo[4,5-d][1,3]dithiole-5-thione |
Safety and Handling
TSCA : No