accessibility menu, dialog, popup

UserName

CAS RN 202126-48-5

IUPAC Name: 5-propyl-2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiole-2-thione
Synonyms: 5-propyl-5H-[1,3]dithiolo[4,5-d][1,3]dithiole-2-thione
Molecular Weight (g/mol): 252.44
Molecular Formula: C7H8S5
InChi Key: QVGTUNLOIXSBRJ-UHFFFAOYSA-N
SMILES: CCCC1SC2=C(S1)SC(=S)S2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results