missing translation for 'onlineSavingsMsg'
Learn More

5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione 93.0+%, TCI America™

Catalog No. p-7135185
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1635250MG 250 mg
P16351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1635250MG Supplier TCI America Supplier No. P1635250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 202126-48-5
Molecular Formula C7H8S5
Molecular Weight (g/mol) 252.441
InChI Key QVGTUNLOIXSBRJ-UHFFFAOYSA-N
PubChem CID 44629778
IUPAC Name 2-propyl-[1,3]dithiolo[4,5-d][1,3]dithiole-5-thione
SMILES CCCC1SC2=C(S1)SC(=S)S2

Specifications

Melting Point 51°C
Quantity 250 mg
Formula Weight 252.44
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione