missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine, 98%

Catalog No. AAJ6513603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6513603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6513603 Supplier Thermo Scientific Chemicals Supplier No. J6513603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93778-57-5 | C31H30N4O6 | 554.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93778-57-5
Molecular Formula C31H30N4O6
Molecular Weight (g/mol) 554.60
MDL Number MFCD00057879
InChI Key IYNGMVFRUKBGNM-OQAPVCBANA-N
Synonym 5'-o-4,4'-dimethoxytrityl-2'-deoxyinosine, dmt-di, 2'-deoxy-5'-o-dmt-inosine, 9-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxyoxolan-2-yl purin-6-ol, inosine,5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy, 2'-deoxy-5'-o-dimethoxytrityl-inosine, 2'-deoxy-5'-ortho-dmt-inosine, 9-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxytetrahydrofuran-2-yl-9h-purin-6-ol, 9-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxyoxolan-2-yl-1h-purin-6-one
PubChem CID 15277171
IUPAC Name 9-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-3H-purin-6-one
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Color White to Yellow
Quantity 1 g
Formula Weight 554.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine
EINECSNumber 298-195-2
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only