accessibility menu, dialog, popup

UserName

CAS RN 93778-57-5

IUPAC Name: 9-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 9-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-1H-purin-6-one
Molecular Weight (g/mol): 554.6
Molecular Formula: C31H30N4O6
InChi Key: IYNGMVFRUKBGNM-OQAPVCBANA-N
SMILES: COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results