missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2,3-dicarboxylic Anhydride 97.0+%, TCI America™

Supplier:  TCI America N0331500G

 View more versions of this product

Catalog No. N0331500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Norbornene-2,3-dicarboxylic Anhydride
826-62-0
500 g
C9H8O3
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Anhydride
C1C2C=CC1C3C2C(=O)OC3=O
13223
≥97.0% (T)
165°C
White
Crystal-Powder at 20°C
MFCD00069013
KNDQHSIWLOJIGP-UHFFFAOYSA-N
164.16
164.16

Chemical Identifiers

826-62-0
164.16
KNDQHSIWLOJIGP-UHFFFAOYSA-N
13223
C9H8O3
MFCD00069013
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Anhydride
C1C2C=CC1C3C2C(=O)OC3=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0620

RTECSNumber : DT5600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions