missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2,3-dicarboxylic Anhydride 97.0+%, TCI America™

Catalog No. p-7133753
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N033125G 25 g
N0331500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N033125G Supplier TCI America Supplier No. N033125G
Only null left

Chemical Identifiers

CAS 826-62-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00069013
InChI Key KNDQHSIWLOJIGP-UHFFFAOYSA-N
Synonym Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Anhydride
PubChem CID 13223
SMILES C1C2C=CC1C3C2C(=O)OC3=O

Specifications

Melting Point 165°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 164.16
Percent Purity ≥97.0% (T)
Chemical Name or Material 5-Norbornene-2,3-dicarboxylic Anhydride
EINECSNumber (4)-0620
RTECSNumber DT5600000
TSCA Yes