Learn More
5-Nitrosalicylic acid, 98%
CAS: 96-97-9 | C7H5NO5 | 183.119 g/mol
Supplier: Thermo Scientific Chemicals A1301006
Description
5-Nitrosalicylic acid is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic, pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Nitrosalicylic acid is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic, pharmaceutical intermediate.
Solubility
Soluble in water 1g/1475ml .
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
Specifications
| 5-Nitrosalicylic acid | |
| 228°C to 236°C | |
| C7H5NO5 | |
| 5 g | |
| 14,6632 | |
| Soluble in water 1g/1475ml . | |
| C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)O | |
| 7318 | |
| 183.12 |
| 96-97-9 | |
| 1.65 | |
| MFCD00007338 | |
| 2213719 | |
| 5-nitrosalicylic acid, 2-hydroxy-5-nitrobenzoic acid, anilotic acid, benzoic acid, 2-hydroxy-5-nitro, 5-nitro-2-hydroxybenzoic acid, unii-82l9g7fyz3, 2-hydroxy-5-nitro-benzoic acid, 5-nitro-salicylic acid, salicylic acid, 5-nitro, 5-nitro salicylic acid | |
| PPDRLQLKHRZIJC-UHFFFAOYSA-N | |
| 183.119 | |
| CHEBI:61281 | |
| 98% |
Chemical Identifiers
| 96-97-9 | |
| 183.119 | |
| PPDRLQLKHRZIJC-UHFFFAOYSA-N | |
| 7318 | |
| C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)O |
| C7H5NO5 | |
| MFCD00007338 | |
| 5-nitrosalicylic acid, 2-hydroxy-5-nitrobenzoic acid, anilotic acid, benzoic acid, 2-hydroxy-5-nitro, 5-nitro-2-hydroxybenzoic acid, unii-82l9g7fyz3, 2-hydroxy-5-nitro-benzoic acid, 5-nitro-salicylic acid, salicylic acid, 5-nitro, 5-nitro salicylic acid | |
| CHEBI:61281 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-548-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only