missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroindole-2-carboxylic acid, 96%

Catalog No. p-7057095
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6644103 1 g
AAH6644106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6644103 Supplier Thermo Scientific Chemicals Supplier No. H6644103

CAS: 16730-20-4 | C9H6N2O4 | 206.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16730-20-4
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.157
MDL Number MFCD00236372
InChI Key LHFOJSCXLFKDIR-UHFFFAOYSA-N
Synonym 5-nitroindole-2-carboxylic acid, 5-nitro-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-nitro, 5-nitro-2-indolecarboxylic acid, 1h-indole-2-carboxylicacid, 5-nitro, 5-nitroindole-2-carboxylicacid, pubchem1710, zlchem 1340, 5-nitro indole-2-carboxylic acid
PubChem CID 351291
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C=C(N2)C(=O)O

Specifications

Quantity 1 g
Beilstein 203744
Formula Weight 206.16
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Nitroindole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only