missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroguaiacol Sodium Salt 98.0+%, TCI America™

Catalog No. N020525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N020525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N020525G Supplier TCI America Supplier No. N020525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67233-85-6
Molecular Formula C7H6NNaO4
Molecular Weight (g/mol) 191.12
MDL Number MFCD00070570
InChI Key KBRKFTKQRMYINW-UHFFFAOYSA-M
Synonym 2-Methoxy-5-nitrophenol Sodium Salt
PubChem CID 3017748
IUPAC Name sodium 2-methoxy-5-nitrobenzen-1-olate
SMILES [Na+].COC1=CC=C(C=C1[O-])[N+]([O-])=O

Specifications

Color Red
Quantity 25 g
Formula Weight 191.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroguaiacol Sodium Salt
EINECSNumber (9)-0971
RTECSNumber SL7807000
TSCA No