accessibility menu, dialog, popup

UserName

NaH3COONOO

CAS RN 67233-85-6

IUPAC Name: sodium 2-methoxy-5-nitrobenzen-1-olate
Synonyms: sodium 5-nitroguaiacolate
Molecular Weight (g/mol): 191.12
Molecular Formula: C7H6NNaO4
InChi Key: KBRKFTKQRMYINW-UHFFFAOYSA-M
SMILES: [Na+].COC1=CC=C(C=C1[O-])[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results