missing translation for 'onlineSavingsMsg'
Learn More

(5-Methylthien-2-yl)methylamine hydrochloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 171661-55-5
Molecular Formula C6H10ClNS
Molecular Weight (g/mol) 163.663
InChI Key COTMVCYERKXKDX-UHFFFAOYSA-N
Synonym 5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthien-2-yl methylamine hydrochloride, 5-methylthien-2-yl methylamine hcl, 1-5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthiophen-2-yl methanaminehydrochloride, 5-methylthien-2-yl methylamine-hcl, 5-methyl-2-thienyl methylamine, chloride, 2-aminomethyl-5-methylthiophene hydrochloride, 2-thiophenemethanamine, 5-methyl-, hydrochloride, 2-thiophenemethanamine,5-methyl-,hydrochloride 1:1
PubChem CID 24229725
IUPAC Name (5-methylthiophen-2-yl)methanamine;hydrochloride
SMILES CC1=CC=C(S1)CN.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC65063CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC65063DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC65063DE
View Documents Promotion Details
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

171661-55-5
163.663
5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthien-2-yl methylamine hydrochloride, 5-methylthien-2-yl methylamine hcl, 1-5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthiophen-2-yl methanaminehydrochloride, 5-methylthien-2-yl methylamine-hcl, 5-methyl-2-thienyl methylamine, chloride, 2-aminomethyl-5-methylthiophene hydrochloride, 2-thiophenemethanamine, 5-methyl-, hydrochloride, 2-thiophenemethanamine,5-methyl-,hydrochloride 1:1
(5-methylthiophen-2-yl)methanamine;hydrochloride
C6H10ClNS
COTMVCYERKXKDX-UHFFFAOYSA-N
24229725
CC1=CC=C(S1)CN.Cl

Specifications

171661-55-5
C6H10ClNS
COTMVCYERKXKDX-UHFFFAOYSA-N
(5-methylthiophen-2-yl)methanamine;hydrochloride
24229725
97%
97%
5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthien-2-yl methylamine hydrochloride, 5-methylthien-2-yl methylamine hcl, 1-5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthiophen-2-yl methanaminehydrochloride, 5-methylthien-2-yl methylamine-hcl, 5-methyl-2-thienyl methylamine, chloride, 2-aminomethyl-5-methylthiophene hydrochloride, 2-thiophenemethanamine, 5-methyl-, hydrochloride, 2-thiophenemethanamine,5-methyl-,hydrochloride 1:1
CC1=CC=C(S1)CN.Cl
163.663
163.67
(5-methylthien-2-yl)methylamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents