missing translation for 'onlineSavingsMsg'
Learn More

(5-Methylthien-2-yl)methylamine hydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC65063DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC65063DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

171661-55-5
163.663
5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthien-2-yl methylamine hydrochloride, 5-methylthien-2-yl methylamine hcl, 1-5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthiophen-2-yl methanaminehydrochloride, 5-methylthien-2-yl methylamine-hcl, 5-methyl-2-thienyl methylamine, chloride, 2-aminomethyl-5-methylthiophene hydrochloride, 2-thiophenemethanamine, 5-methyl-, hydrochloride, 2-thiophenemethanamine,5-methyl-,hydrochloride 1:1
(5-methylthiophen-2-yl)methanamine;hydrochloride
C6H10ClNS
COTMVCYERKXKDX-UHFFFAOYSA-N
24229725
CC1=CC=C(S1)CN.Cl

Specifications

(5-methylthien-2-yl)methylamine hydrochloride
97%
C6H10ClNS
5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthien-2-yl methylamine hydrochloride, 5-methylthien-2-yl methylamine hcl, 1-5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthiophen-2-yl methanaminehydrochloride, 5-methylthien-2-yl methylamine-hcl, 5-methyl-2-thienyl methylamine, chloride, 2-aminomethyl-5-methylthiophene hydrochloride, 2-thiophenemethanamine, 5-methyl-, hydrochloride, 2-thiophenemethanamine,5-methyl-,hydrochloride 1:1
CC1=CC=C(S1)CN.Cl
163.663
163.67
171661-55-5
Amber Glass Bottle
1g
COTMVCYERKXKDX-UHFFFAOYSA-N
(5-methylthiophen-2-yl)methanamine;hydrochloride
24229725
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions