missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5-Methylthien-2-yl)methylamine hydrochloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC65063CB
| Quantity | 250mg |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 171661-55-5 | |
| 163.663 | |
| 5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthien-2-yl methylamine hydrochloride, 5-methylthien-2-yl methylamine hcl, 1-5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthiophen-2-yl methanaminehydrochloride, 5-methylthien-2-yl methylamine-hcl, 5-methyl-2-thienyl methylamine, chloride, 2-aminomethyl-5-methylthiophene hydrochloride, 2-thiophenemethanamine, 5-methyl-, hydrochloride, 2-thiophenemethanamine,5-methyl-,hydrochloride 1:1 | |
| (5-methylthiophen-2-yl)methanamine;hydrochloride |
| C6H10ClNS | |
| COTMVCYERKXKDX-UHFFFAOYSA-N | |
| 24229725 | |
| CC1=CC=C(S1)CN.Cl |
Specifications
| (5-methylthien-2-yl)methylamine hydrochloride | |
| 97% | |
| C6H10ClNS | |
| 5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthien-2-yl methylamine hydrochloride, 5-methylthien-2-yl methylamine hcl, 1-5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthiophen-2-yl methanaminehydrochloride, 5-methylthien-2-yl methylamine-hcl, 5-methyl-2-thienyl methylamine, chloride, 2-aminomethyl-5-methylthiophene hydrochloride, 2-thiophenemethanamine, 5-methyl-, hydrochloride, 2-thiophenemethanamine,5-methyl-,hydrochloride 1:1 | |
| CC1=CC=C(S1)CN.Cl | |
| 163.663 | |
| 163.67 |
| 171661-55-5 | |
| Amber Glass Bottle | |
| 250mg | |
| COTMVCYERKXKDX-UHFFFAOYSA-N | |
| (5-methylthiophen-2-yl)methanamine;hydrochloride | |
| 24229725 | |
| 97% |
Safety and Handling
GHS Signal Word: Warning