missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylsalicylic Acid 98.0+%, TCI America™
$77.00 - $231.02
Chemical Identifiers
| CAS | 89-56-5 |
|---|---|
| Molecular Formula | C8H8O3 |
| Molecular Weight (g/mol) | 152.149 |
| MDL Number | MFCD00002461 |
| InChI Key | DLGBEGBHXSAQOC-UHFFFAOYSA-N |
| Synonym | 5-methylsalicylic acid, p-cresotic acid, p-cresotinic acid, 6-hydroxy-m-toluic acid, p-homosalicylic acid, 2,5-cresotic acid, benzoic acid, 2-hydroxy-5-methyl, 5-methyl-2-hydroxybenzoic acid, 6-hydroxy-3-methylbenzoic acid, alpha-cresotinic acid |
| PubChem CID | 6973 |
| IUPAC Name | 2-hydroxy-5-methylbenzoic acid |
| SMILES | CC1=CC(=C(C=C1)O)C(=O)O |
Chemical Identifiers
| 89-56-5 | |
| 152.149 | |
| DLGBEGBHXSAQOC-UHFFFAOYSA-N | |
| 6973 | |
| CC1=CC(=C(C=C1)O)C(=O)O |
| C8H8O3 | |
| MFCD00002461 | |
| 5-methylsalicylic acid, p-cresotic acid, p-cresotinic acid, 6-hydroxy-m-toluic acid, p-homosalicylic acid, 2,5-cresotic acid, benzoic acid, 2-hydroxy-5-methyl, 5-methyl-2-hydroxybenzoic acid, 6-hydroxy-3-methylbenzoic acid, alpha-cresotinic acid | |
| 2-hydroxy-5-methylbenzoic acid |
Specifications
| 89-56-5 | |
| White-Yellow | |
| MFCD00002461 | |
| 5-methylsalicylic acid, p-cresotic acid, p-cresotinic acid, 6-hydroxy-m-toluic acid, p-homosalicylic acid, 2,5-cresotic acid, benzoic acid, 2-hydroxy-5-methyl, 5-methyl-2-hydroxybenzoic acid, 6-hydroxy-3-methylbenzoic acid, alpha-cresotinic acid | |
| CC1=CC(=C(C=C1)O)C(=O)O | |
| 152.149 | |
| 152.15 | |
| Crystalline Powder |
| 151°C | |
| C8H8O3 | |
| 5 g | |
| DLGBEGBHXSAQOC-UHFFFAOYSA-N | |
| 2-hydroxy-5-methylbenzoic acid | |
| 6973 | |
| ≥98.0% (T) | |
| 5-Methylsalicylic Acid |
Safety and Handling
RTECSNumber : GP3920200
TSCA : No