missing translation for 'onlineSavingsMsg'
Learn More

5-Methylsalicylic acid, 98%

Catalog No. AAB2004006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2004006 5 g
AAB2004014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2004006 Supplier Thermo Scientific Chemicals Supplier No. B2004006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 89-56-5 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-56-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002461
InChI Key DLGBEGBHXSAQOC-UHFFFAOYSA-N
Synonym 5-methylsalicylic acid, p-cresotic acid, p-cresotinic acid, 6-hydroxy-m-toluic acid, p-homosalicylic acid, 2,5-cresotic acid, benzoic acid, 2-hydroxy-5-methyl, 5-methyl-2-hydroxybenzoic acid, 6-hydroxy-3-methylbenzoic acid, alpha-cresotinic acid
PubChem CID 6973
IUPAC Name 2-hydroxy-5-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 151°C to 154°C
Odor Odorless
Quantity 5 g
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 5-Methylsalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber GP3920200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only