missing translation for 'onlineSavingsMsg'
Learn More

5-Methylresorcinol Monohydrate 98.0+%, TCI America™

Supplier:  TCI America M042625G

 View more versions of this product

Catalog No. M042625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Methylresorcinol Monohydrate
59°C
MFCD00149092
5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate
CC1=CC(=CC(=C1)O)O.O
142.154
124.14
Crystalline Powder
6153-39-5
C7H10O3
25 g
NBKPNAMTHBIMLA-UHFFFAOYSA-N
5-methylbenzene-1,3-diol;hydrate
3083941
≥98.0% (GC)

Chemical Identifiers

6153-39-5
142.154
NBKPNAMTHBIMLA-UHFFFAOYSA-N
3083941
CC1=CC(=CC(=C1)O)O.O
C7H10O3
MFCD00149092
5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate
5-methylbenzene-1,3-diol;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0057

RTECSNumber : VH2100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions