missing translation for 'onlineSavingsMsg'
Learn More

5-Methylresorcinol Monohydrate 98.0+%, TCI America™

Catalog No. p-7132163
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
M04265G 5 g
M042625G 25 g
2 options

Catalog No. M04265G

Supplier: TCI America M04265G

Only null left

Chemical Identifiers

CAS 6153-39-5
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00149092
InChI Key NBKPNAMTHBIMLA-UHFFFAOYSA-N
Synonym 5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate
PubChem CID 3083941
IUPAC Name 5-methylbenzene-1,3-diol;hydrate
SMILES CC1=CC(=CC(=C1)O)O.O

Specifications

Melting Point 59°C
Quantity 5 g
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylresorcinol Monohydrate
EINECSNumber (4)-0057
RTECSNumber VH2100000
TSCA Yes