missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylresorcinol Anhydrous 98.0+%, TCI America™
Supplier: TCI America M123525G
Specifications
| 5-Methylresorcinol Anhydrous | |
| 110°C | |
| C7H8O2 | |
| 25 g | |
| OIPPWFOQEKKFEE-UHFFFAOYSA-N | |
| 5-methylbenzene-1,3-diol | |
| 10436 | |
| 124.14 | |
| Crystalline Powder |
| 504-15-4 | |
| 147°C | |
| MFCD00002291 | |
| orcinol, 3,5-dihydroxytoluene, 5-methylresorcinol, 1,3-dihydroxy-5-methylbenzene, orcin, 5-methyl-1,3-benzenediol, 5-methylresorcin, 3-hydroxy-5-methylphenol, 3,5-toluenediol, 1,3-benzenediol, 5-methyl | |
| CC1=CC(O)=CC(O)=C1 | |
| 124.14 | |
| CHEBI:16536 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 504-15-4 | |
| 124.14 | |
| OIPPWFOQEKKFEE-UHFFFAOYSA-N | |
| 10436 | |
| 5-methylbenzene-1,3-diol |
| C7H8O2 | |
| MFCD00002291 | |
| orcinol, 3,5-dihydroxytoluene, 5-methylresorcinol, 1,3-dihydroxy-5-methylbenzene, orcin, 5-methyl-1,3-benzenediol, 5-methylresorcin, 3-hydroxy-5-methylphenol, 3,5-toluenediol, 1,3-benzenediol, 5-methyl | |
| CHEBI:16536 | |
| CC1=CC(O)=CC(O)=C1 |
Safety and Handling
EINECSNumber : (4)-0057
RTECSNumber : VH2100000
TSCA : Yes