accessibility menu, dialog, popup

UserName

CAS RN 504-15-4

IUPAC Name: 5-methylbenzene-1,3-diol
Synonyms: orcinol orcinol, 14c-labeled cpd orcinol, 14C-labeled cpd orcin 5-methylresorcinol orcinol orzin 14c-labeled cpd orcinol orcinol, 5-methylresorcinol 14C-labeled cpd orcinol
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: OIPPWFOQEKKFEE-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(O)=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)