missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylisoxazole 95.0+%, TCI America™
$130.48 - $434.32
Chemical Identifiers
| CAS | 5765-44-6 |
|---|---|
| Molecular Formula | C4H5NO |
| Molecular Weight (g/mol) | 83.09 |
| MDL Number | MFCD00003152 |
| InChI Key | AGQOIYCTCOEHGR-UHFFFAOYSA-N |
| Synonym | 5-methylisoxazole, isoxazole, 5-methyl, unii-yc2p4ibf1e, yc2p4ibf1e, 5-methyl isoxazole, 5-methylisoxazol, 5-methyl-isoxazole, pubchem8634, acmc-1arbg |
| PubChem CID | 79833 |
| IUPAC Name | 5-methyl-1,2-oxazole |
| SMILES | CC1=CC=NO1 |
Chemical Identifiers
| 5765-44-6 | |
| 83.09 | |
| AGQOIYCTCOEHGR-UHFFFAOYSA-N | |
| 79833 | |
| CC1=CC=NO1 |
| C4H5NO | |
| MFCD00003152 | |
| 5-methylisoxazole, isoxazole, 5-methyl, unii-yc2p4ibf1e, yc2p4ibf1e, 5-methyl isoxazole, 5-methylisoxazol, 5-methyl-isoxazole, pubchem8634, acmc-1arbg | |
| 5-methyl-1,2-oxazole |
Specifications
| 5765-44-6 | |
| 122°C | |
| MFCD00003152 | |
| 1993 | |
| AGQOIYCTCOEHGR-UHFFFAOYSA-N | |
| 5-methyl-1,2-oxazole | |
| 79833 | |
| ≥95.0% (GC) | |
| 5-Methylisoxazole |
| Yellow | |
| C4H5NO | |
| 5 mL | |
| 5-methylisoxazole, isoxazole, 5-methyl, unii-yc2p4ibf1e, yc2p4ibf1e, 5-methyl isoxazole, 5-methylisoxazol, 5-methyl-isoxazole, pubchem8634, acmc-1arbg | |
| CC1=CC=NO1 | |
| 83.09 | |
| 83.09 | |
| Liquid |
Safety and Handling
TSCA : Yes