CAS RN 5765-44-6
CAS RN 5765-44-6
IUPAC Name:
5-methyl-1,2-oxazole
Synonyms:
isoxazole, 5-methyl-
Molecular Weight (g/mol):
83.09
Molecular Formula:
C4H5NO
InChi Key:
AGQOIYCTCOEHGR-UHFFFAOYSA-N
SMILES:
CC1=CC=NO1
1
–
1
of
1
results
1
5-Methylisoxazole 95.0+%, TCI America™
CAS: 5765-44-6 Molecular Formula: C4H5NO Molecular Weight (g/mol): 83.09 MDL Number: MFCD00003152 InChI Key: AGQOIYCTCOEHGR-UHFFFAOYSA-N Synonym: 5-methylisoxazole,isoxazole, 5-methyl,unii-yc2p4ibf1e,yc2p4ibf1e,5-methyl isoxazole,5-methylisoxazol,5-methyl-isoxazole,pubchem8634,acmc-1arbg PubChem CID: 79833 IUPAC Name: 5-methyl-1,2-oxazole SMILES: CC1=CC=NO1