missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylisoxazole 95.0+%, TCI America™
Supplier: TCI America M081325ML
Specifications
| 5-Methylisoxazole | |
| Yellow | |
| C4H5NO | |
| 25 mL | |
| 5-methylisoxazole, isoxazole, 5-methyl, unii-yc2p4ibf1e, yc2p4ibf1e, 5-methyl isoxazole, 5-methylisoxazol, 5-methyl-isoxazole, pubchem8634, acmc-1arbg | |
| CC1=CC=NO1 | |
| 83.09 | |
| 83.09 | |
| Liquid |
| 5765-44-6 | |
| 122°C | |
| MFCD00003152 | |
| 1993 | |
| AGQOIYCTCOEHGR-UHFFFAOYSA-N | |
| 5-methyl-1,2-oxazole | |
| 79833 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 5765-44-6 | |
| 83.09 | |
| AGQOIYCTCOEHGR-UHFFFAOYSA-N | |
| 79833 | |
| CC1=CC=NO1 |
| C4H5NO | |
| MFCD00003152 | |
| 5-methylisoxazole, isoxazole, 5-methyl, unii-yc2p4ibf1e, yc2p4ibf1e, 5-methyl isoxazole, 5-methylisoxazol, 5-methyl-isoxazole, pubchem8634, acmc-1arbg | |
| 5-methyl-1,2-oxazole |
Safety and Handling
TSCA : Yes