missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylhexanoic Acid 98.0+%, TCI America™
$74.05 - $307.57
Chemical Identifiers
| CAS | 628-46-6 |
|---|---|
| Molecular Formula | C7H14O2 |
| Molecular Weight (g/mol) | 130.19 |
| MDL Number | MFCD00036494 |
| InChI Key | MHPUGCYGQWGLJL-UHFFFAOYSA-N |
| Synonym | isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2 |
| PubChem CID | 12344 |
| IUPAC Name | 5-methylhexanoic acid |
| SMILES | CC(C)CCCC(O)=O |
Chemical Identifiers
| C7H14O2 | |
| MFCD00036494 | |
| isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2 | |
| 5-methylhexanoic acid |
Specifications
| 628-46-6 | |
| 216°C | |
| MFCD00036494 | |
| isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2 | |
| CC(C)CCCC(O)=O | |
| 130.19 | |
| 130.19 | |
| Liquid |
| Yellow | |
| C7H14O2 | |
| 3265 | |
| MHPUGCYGQWGLJL-UHFFFAOYSA-N | |
| 5-methylhexanoic acid | |
| 12344 | |
| ≥98.0% (T,GC) | |
| 5-Methylhexanoic Acid |
Safety and Handling
RTECSNumber : MO8400900
TSCA : Yes