accessibility menu, dialog, popup

UserName

CAS RN 628-46-6

IUPAC Name: 5-methylhexanoic acid
Synonyms: 5-methylhexanoic acid
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: MHPUGCYGQWGLJL-UHFFFAOYSA-N
SMILES: CC(C)CCCC(O)=O
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)