missing translation for 'onlineSavingsMsg'
Learn More

5-Methylhexanoic Acid 98.0+%, TCI America™

Catalog No. M11425ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11425ML 5 mL
M114225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11425ML Supplier TCI America Supplier No. M11425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-46-6
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00036494
InChI Key MHPUGCYGQWGLJL-UHFFFAOYSA-N
Synonym isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2
PubChem CID 12344
IUPAC Name 5-methylhexanoic acid
SMILES CC(C)CCCC(O)=O

Specifications

Color Yellow
Boiling Point 216°C
Quantity 5 mL
UN Number 3265
Formula Weight 130.19
Percent Purity ≥98.0% (T,GC)
Physical Form Liquid
Chemical Name or Material 5-Methylhexanoic Acid
RTECSNumber MO8400900
TSCA Yes