missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylbenzoxazole 98.0+%, TCI America™
Supplier: TCI America M168825G
Specifications
| 5-Methylbenzoxazole | |
| 45°C | |
| 96°C | |
| MFCD00216938 | |
| 5-methylbenzoxazole, 5-methylbenzo d oxazole, 5-methylbenzooxazole, 5-methyl-benzoxazole, pubchem8638, acmc-1bprw, 5-methylbenzo d oxazole;5-methylbenzo d oxazole | |
| CC1=CC=C2OC=NC2=C1 | |
| 133.15 | |
| 133.15 | |
| Crystalline Powder |
| 10531-78-9 | |
| White-Yellow | |
| C8H7NO | |
| 25 g | |
| UBIAVBGIRDRQLD-UHFFFAOYSA-N | |
| 5-methyl-1,3-benzoxazole | |
| 2724970 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 10531-78-9 | |
| 133.15 | |
| UBIAVBGIRDRQLD-UHFFFAOYSA-N | |
| 2724970 | |
| CC1=CC=C2OC=NC2=C1 |
| C8H7NO | |
| MFCD00216938 | |
| 5-methylbenzoxazole, 5-methylbenzo d oxazole, 5-methylbenzooxazole, 5-methyl-benzoxazole, pubchem8638, acmc-1bprw, 5-methylbenzo d oxazole;5-methylbenzo d oxazole | |
| 5-methyl-1,3-benzoxazole |
Safety and Handling
TSCA : No