accessibility menu, dialog, popup

UserName

CAS RN 10531-78-9

IUPAC Name: 5-methyl-1,3-benzoxazole
Synonyms: 5-methyl-1,3-benzoxazole
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: UBIAVBGIRDRQLD-UHFFFAOYSA-N
SMILES: CC1=CC=C2OC=NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results