missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzoxazole 98.0+%, TCI America™

Catalog No. p-7132809
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16885G 5 g
M168825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16885G Supplier TCI America Supplier No. M16885G
Only null left

Chemical Identifiers

CAS 10531-78-9
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00216938
InChI Key UBIAVBGIRDRQLD-UHFFFAOYSA-N
Synonym 5-methylbenzoxazole, 5-methylbenzo d oxazole, 5-methylbenzooxazole, 5-methyl-benzoxazole, pubchem8638, acmc-1bprw, 5-methylbenzo d oxazole;5-methylbenzo d oxazole
PubChem CID 2724970
IUPAC Name 5-methyl-1,3-benzoxazole
SMILES CC1=CC=C2OC=NC2=C1

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylbenzoxazole