missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-nitrophenol, 97%

CAS: 700-38-9 | C7H7NO3 | 153.14 g/mol

$90.09 - $90.09

Chemical Identifiers

CAS 700-38-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007111
InChI Key NQXUSSVLFOBRSE-UHFFFAOYSA-N
Synonym 6-nitro-m-cresol, phenol, 5-methyl-2-nitro, 3-hydroxy-4-nitrotoluene, 6-nitro-3-cresol, 3-methyl-6-nitrophenol, m-cresol, 6-nitro, 5-methyl-2-nitro-phenol, unii-z0b5eo9752, 6-nitro-m-creso
PubChem CID 12788
IUPAC Name 5-methyl-2-nitrophenol
SMILES CC1=CC=C(C(O)=C1)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC160150250
SDS View Documents
Thermo Scientific Chemicals
160150250
25 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

700-38-9
153.14
NQXUSSVLFOBRSE-UHFFFAOYSA-N
12788
CC1=CC=C(C(O)=C1)[N+]([O-])=O
C7H7NO3
MFCD00007111
6-nitro-m-cresol, phenol, 5-methyl-2-nitro, 3-hydroxy-4-nitrotoluene, 6-nitro-3-cresol, 3-methyl-6-nitrophenol, m-cresol, 6-nitro, 5-methyl-2-nitro-phenol, unii-z0b5eo9752, 6-nitro-m-creso
5-methyl-2-nitrophenol

Specifications

700-38-9
100.0
Green to Yellow
Authentic
Glass bottle
CH3C6H3(NO2)OH
25 g
6-nitro-m-cresol, phenol, 5-methyl-2-nitro, 3-hydroxy-4-nitrotoluene, 6-nitro-3-cresol, 3-methyl-6-nitrophenol, m-cresol, 6-nitro, 5-methyl-2-nitro-phenol, unii-z0b5eo9752, 6-nitro-m-creso
CC1=CC=C(C(O)=C1)[N+]([O-])=O
153.14
153.14
Crystalline Chunks
96.0
53°C to 56°C
109°C
96% min. (GC)
C7H7NO3
MFCD00007111
06, 385
NQXUSSVLFOBRSE-UHFFFAOYSA-N
5-methyl-2-nitrophenol
12788
97%
5-Methyl-2-nitrophenol, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 211-843-0

RTECSNumber : SM1130950

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only