missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-nitrobenzoic acid, 98+%

Catalog No. AAL0659406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0659406 5 g
AAL0659414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0659406 Supplier Thermo Scientific Chemicals Supplier No. L0659406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3113-72-2 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3113-72-2
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007368
InChI Key QRRSIFNWHCKMSW-UHFFFAOYSA-N
Synonym 5-methyl-2-nitrobenzoic acid, 2-nitro-5-methylbenzoic acid, benzoic acid, 5-methyl-2-nitro, 3-methyl-6-nitrobenzoic acid, 6-nitro-m-toluic acid, m-toluic acid, 6-nitro, 6-nitro-p-toluic acid, 5-methyl-2-nitrobenzoicacid, 5-methyl-2-nitro-benzoic acid, benzoic acid,5-methyl-2-nitro
PubChem CID 18371
SMILES CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 133°C to 137°C
Quantity 5 g
Beilstein 1959031
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 5-Methyl-2-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-481-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only