accessibility menu, dialog, popup

UserName

CH3HOONOO

CAS RN 3113-72-2

IUPAC Name: 5-methyl-2-nitrobenzoic acid
Synonyms: benzoic acid,5-methyl-2-nitro
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: QRRSIFNWHCKMSW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(=C1)C(O)=O)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)