missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methylaniline 98.0+%, TCI America™

Catalog No. M30091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30091G 1 g
M30095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30091G Supplier TCI America Supplier No. M30091G

Chemical Identifiers

CAS 50868-72-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00075057
InChI Key RPJXLEZOFUNGNZ-UHFFFAOYSA-N
Synonym 5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic
PubChem CID 99500
IUPAC Name 5-methoxy-2-methylaniline
SMILES COC1=CC=C(C)C(N)=C1

Specifications

Melting Point 44°C
Boiling Point 253°C
Quantity 1 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxy-2-methylaniline
EINECSNumber (3)-0614
TSCA No