accessibility menu, dialog, popup

UserName

H3COCH3H2N

CAS RN 50868-72-9

IUPAC Name: 5-methoxy-2-methylaniline
Synonyms: 2-methyl-5-ethoxyaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: RPJXLEZOFUNGNZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C(N)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)