missing translation for 'onlineSavingsMsg'
Learn More

5-Isopropyl-2-methoxybenzeneboronic acid, 98+%

Catalog No. AAL1746003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1746003 1 g
AAL1746006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1746003 Supplier Thermo Scientific Chemicals Supplier No. L1746003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 216393-63-4 | C10H15BO3 | 194.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 216393-63-4
Molecular Formula C10H15BO3
Molecular Weight (g/mol) 194.037
MDL Number MFCD01318154
InChI Key UTKAEAGLVJFBMK-UHFFFAOYSA-N
Synonym 5-isopropyl-2-methoxyphenylboronic acid, 5-isopropyl-2-methoxyphenyl boronic acid, 5-isopropyl-2-methoxybenzeneboronic acid, boronic acid, 2-methoxy-5-1-methylethyl phenyl, 2-methoxy-5-propan-2-ylphenyl boronic acid, 2-methoxy-5-1-methylethyl phenyl boronic acid, 2-methoxy-5-propan-2-yl phenyl boronic acid, acmc-1cp8q, 2-methoxy-5-propan-2-yl-phenyl boronic acid, 5-isopropyl-2-methoxyphenyl boronicacid
PubChem CID 4589190
IUPAC Name (2-methoxy-5-propan-2-ylphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)C(C)C)OC)(O)O

Specifications

Melting Point 85°C to 87°C
Quantity 1 g
Beilstein 9193266
Formula Weight 194.04
Percent Purity ≥98%
Chemical Name or Material 5-Isopropyl-2-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only