missing translation for 'onlineSavingsMsg'
Learn More

5-Iodouridine, 96%

CAS: 1024-99-3 | C9H11IN2O6 | 370.099 g/mol

$169.82 - $630.22

Chemical Identifiers

CAS 1024-99-3
Molecular Formula C9H11IN2O6
Molecular Weight (g/mol) 370.099
MDL Number MFCD00006532
InChI Key RKSLVDIXBGWPIS-UAKXSSHOSA-N
Synonym 5-iodouridine, uridine, 5-iodo, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidine-2,4 1h,3h-dione, 5-iodo-uridine, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-iodo-3h-pyrimidine-2,4-dione, 5-i-uridine, 2,4-dihydroxy-5-iodo-1-beta-d-ribofuranosylpyrimidine, ksc179a6b, uridine, 5-iodo-8ci 9ci
PubChem CID 1268108
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
SMILES C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6225903
View Documents
Thermo Scientific Chemicals
J6225903
1 g
Each for $169.82
Only null left
 
AAJ6225906
View Documents
Thermo Scientific Chemicals
J6225906
5 g
Each for $630.22
Only null left
 
Description

Description

5-Iodouridine has shown to enhance the effect of gamma irradiation in hamster cells. It is used as catalytic agent, petrochemical additive, used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodouridine has shown to enhance the effect of gamma irradiation in hamster cells. It is used as catalytic agent, petrochemical additive, used in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

1024-99-3
370.099
RKSLVDIXBGWPIS-UAKXSSHOSA-N
1268108
C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)I
C9H11IN2O6
MFCD00006532
5-iodouridine, uridine, 5-iodo, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidine-2,4 1h,3h-dione, 5-iodo-uridine, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-iodo-3h-pyrimidine-2,4-dione, 5-i-uridine, 2,4-dihydroxy-5-iodo-1-beta-d-ribofuranosylpyrimidine, ksc179a6b, uridine, 5-iodo-8ci 9ci
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione

Specifications

1024-99-3
C9H11IN2O6
1 g
5-iodouridine, uridine, 5-iodo, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidine-2,4 1h,3h-dione, 5-iodo-uridine, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-iodo-3h-pyrimidine-2,4-dione, 5-i-uridine, 2,4-dihydroxy-5-iodo-1-beta-d-ribofuranosylpyrimidine, ksc179a6b, uridine, 5-iodo-8ci 9ci
RKSLVDIXBGWPIS-UAKXSSHOSA-N
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
1268108
96%
5-Iodouridine
White
MFCD00006532
33665
Soluble in water.
C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)I
370.099
370.1
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-833-1

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only