Learn More
5-Iodouridine, 96%
CAS: 1024-99-3 | C9H11IN2O6 | 370.099 g/mol
Supplier: Thermo Scientific Chemicals J6225906
Description
5-Iodouridine has shown to enhance the effect of gamma irradiation in hamster cells. It is used as catalytic agent, petrochemical additive, used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Iodouridine has shown to enhance the effect of gamma irradiation in hamster cells. It is used as catalytic agent, petrochemical additive, used in organic synthesis.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications
| 5-Iodouridine | |
| White | |
| MFCD00006532 | |
| 33665 | |
| Soluble in water. | |
| C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)I | |
| 370.099 | |
| 370.1 | |
| Powder |
| 1024-99-3 | |
| C9H11IN2O6 | |
| 5 g | |
| 5-iodouridine, uridine, 5-iodo, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidine-2,4 1h,3h-dione, 5-iodo-uridine, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-iodo-3h-pyrimidine-2,4-dione, 5-i-uridine, 2,4-dihydroxy-5-iodo-1-beta-d-ribofuranosylpyrimidine, ksc179a6b, uridine, 5-iodo-8ci 9ci | |
| RKSLVDIXBGWPIS-UAKXSSHOSA-N | |
| 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione | |
| 1268108 | |
| 96% |
Chemical Identifiers
| 1024-99-3 | |
| 370.099 | |
| RKSLVDIXBGWPIS-UAKXSSHOSA-N | |
| 1268108 | |
| C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)I |
| C9H11IN2O6 | |
| MFCD00006532 | |
| 5-iodouridine, uridine, 5-iodo, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidine-2,4 1h,3h-dione, 5-iodo-uridine, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-5-iodo-3h-pyrimidine-2,4-dione, 5-i-uridine, 2,4-dihydroxy-5-iodo-1-beta-d-ribofuranosylpyrimidine, ksc179a6b, uridine, 5-iodo-8ci 9ci | |
| 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione |
Safety and Handling
EINECSNumber : 213-833-1
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only