missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Iodo-m-terphenyl 98.0+%, TCI America™
$177.08 - $615.14
Chemical Identifiers
| CAS | 87666-86-2 |
|---|---|
| Molecular Formula | C18H13I |
| Molecular Weight (g/mol) | 356.206 |
| MDL Number | MFCD26131291 |
| InChI Key | LICOFADCKUKHJI-UHFFFAOYSA-N |
| Synonym | 1-Iodo-3,5-diphenylbenzene, 5′C-Iodo-1,1′C:3′C,1′C′C-terphenyl |
| PubChem CID | 13067230 |
| IUPAC Name | 1-iodo-3,5-diphenylbenzene |
| SMILES | C1=CC=C(C=C1)C2=CC(=CC(=C2)I)C3=CC=CC=C3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I0969200MG
|
TCI America
I0969200MG |
200 mg |
Each for $177.08
|
|
|||||
|
I09691G
|
TCI America
I09691G |
1 g |
Each for $615.14
|
|
|||||
Chemical Identifiers
| 87666-86-2 | |
| 356.206 | |
| LICOFADCKUKHJI-UHFFFAOYSA-N | |
| 13067230 | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2)I)C3=CC=CC=C3 |
| C18H13I | |
| MFCD26131291 | |
| 1-Iodo-3,5-diphenylbenzene, 5′C-Iodo-1,1′C:3′C,1′C′C-terphenyl | |
| 1-iodo-3,5-diphenylbenzene |
Specifications
| 87666-86-2 | |
| C18H13I | |
| 200 mg | |
| LICOFADCKUKHJI-UHFFFAOYSA-N | |
| 1-iodo-3,5-diphenylbenzene | |
| 13067230 | |
| ≥98.0% (GC) | |
| 5′-Iodo-m-terphenyl |
| 103°C | |
| MFCD26131291 | |
| 1-Iodo-3,5-diphenylbenzene, 5′C-Iodo-1,1′C:3′C,1′C′C-terphenyl | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2)I)C3=CC=CC=C3 | |
| 356.206 | |
| 356.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No