missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Iodo-m-terphenyl 98.0+%, TCI America™
Supplier: TCI America I09691G
Specifications
| 5′-Iodo-m-terphenyl | |
| 103°C | |
| MFCD26131291 | |
| 1-Iodo-3,5-diphenylbenzene, 5′C-Iodo-1,1′C:3′C,1′C′C-terphenyl | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2)I)C3=CC=CC=C3 | |
| 356.206 | |
| 356.21 | |
| Crystalline Powder |
| 87666-86-2 | |
| C18H13I | |
| 1 g | |
| LICOFADCKUKHJI-UHFFFAOYSA-N | |
| 1-iodo-3,5-diphenylbenzene | |
| 13067230 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 87666-86-2 | |
| 356.206 | |
| LICOFADCKUKHJI-UHFFFAOYSA-N | |
| 13067230 | |
| C1=CC=C(C=C1)C2=CC(=CC(=C2)I)C3=CC=CC=C3 |
| C18H13I | |
| MFCD26131291 | |
| 1-Iodo-3,5-diphenylbenzene, 5′C-Iodo-1,1′C:3′C,1′C′C-terphenyl | |
| 1-iodo-3,5-diphenylbenzene |
Safety and Handling
TSCA : No