missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-methoxybenzaldehyde, 97%

Catalog No. p-7057027
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6632003 1 g
AAH6632006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6632003 Supplier Thermo Scientific Chemicals Supplier No. H6632003

CAS: 42298-41-9 | C8H7IO2 | 262.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42298-41-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
MDL Number MFCD04128999
InChI Key PIRKMHAREMCDPZ-UHFFFAOYSA-N
Synonym 2-methoxy-5-iodobenzaldehyde, 5-iodo-2-methoxy-benzaldehyde, pubchem1445, acmc-20am2z, benzaldehyde,5-iodo-2-methoxy, 5-iodo-2-methoxybenzaldehyde
PubChem CID 262240
IUPAC Name 5-iodo-2-methoxybenzaldehyde
SMILES COC1=CC=C(I)C=C1C=O

Specifications

Melting Point 140°C to 146°C
Color White to Yellow
Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 262.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Iodo-2-methoxybenzaldehyde
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only