accessibility menu, dialog, popup

UserName

CAS RN 42298-41-9

IUPAC Name: 5-iodo-2-methoxybenzaldehyde
Synonyms: 5-iodo-2-methoxybenzaldehyde
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: PIRKMHAREMCDPZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(I)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results