missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-nitrobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7130874
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10395G 5 g
H103925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10395G Supplier TCI America Supplier No. H10395G
Only null left

Chemical Identifiers

CAS 42454-06-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007332
InChI Key XLYPHUGUKGMURE-UHFFFAOYSA-N
Synonym 3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
PubChem CID 39211
IUPAC Name 5-hydroxy-2-nitrobenzaldehyde
SMILES OC1=CC=C(C(C=O)=C1)[N+]([O-])=O

Specifications

Melting Point 172°C
Quantity 5 g
Formula Weight 167.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-2-nitrobenzaldehyde
RTECSNumber CU6700000
TSCA No