missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-1-tetralone, 99%

Catalog No. AAA1703406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1703406 5 g
AAA1703414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1703406 Supplier Thermo Scientific Chemicals Supplier No. A1703406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 28315-93-7 | C10H10O2 | 162.188 g/mol

5-Hydroxy-1-tetralone, is used as a metabolite of Levobunolol (L335000) and d-Bunolol (L335010). It is also used as a reagent for the determination of hexoses and oligosaccharides by a fluorescence technique. A reagent that is used for fluorescence determination of enzyme activity. Some of the other applications include as internal standard during HPLC determination of 4-hydroxymephenytoin (4-OH-M) in human urine, as fluorescent labeling reagent during micro detection of glycosphingolipid on TLC plates, in synthesis of 1,2,3,4-tetrahydro-5H-1-benzazepine-quinone derivatives, in synthesis of new chiral oxathiane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxy-1-tetralone, is used as a metabolite of Levobunolol (L335000) and d-Bunolol (L335010). It is also used as a reagent for the determination of hexoses and oligosaccharides by a fluorescence technique. A reagent that is used for fluorescence determination of enzyme activity. Some of the other applications include as internal standard during HPLC determination of 4-hydroxymephenytoin (4-OH-M) in human urine, as fluorescent labeling reagent during micro detection of glycosphingolipid on TLC plates, in synthesis of 1,2,3,4-tetrahydro-5H-1-benzazepine-quinone derivatives, in synthesis of new chiral oxathiane.

Solubility
Soluble in alcohol, ether, benzene and acetic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, acid chlorides, acid anhydrides. Stable under recommended storage conditions.

Chemical Identifiers

CAS 28315-93-7
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00001693
InChI Key YPPZCRZRQHFRBH-UHFFFAOYSA-N
Synonym 5-hydroxy-1-tetralone, 1-tetralon-5-ol, 5-hydroxy-3,4-dihydronaphthalen-1 2h-one, 5-hydroy-1-tetraione, 1 2h-naphthalenone, 3,4-dihydro-5-hydroxy, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 5-hydroxytetralone, 3,4-dihydro-5-hydroxy-1 2h-naphthalenone, 1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one, 5-hydroxy-2,3,4-trihydronaphthalen-1-one
PubChem CID 119921
IUPAC Name 5-hydroxy-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=CC=C2O)C(=O)C1

Specifications

Melting Point 209°C to 211°C
Quantity 5 g
Beilstein 2437410
Solubility Information Soluble in alcohol,ether,benzene and acetic acid.
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material 5-Hydroxy-1-tetralone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 248-958-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only