missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxy-1-indanone 98.0+%, TCI America™
$156.59 - $482.79
Chemical Identifiers
| CAS | 3470-49-3 |
|---|---|
| Molecular Formula | C9H8O2 |
| Molecular Weight (g/mol) | 148.161 |
| MDL Number | MFCD00857527 |
| InChI Key | ZRKQOVXGDIZYDS-UHFFFAOYSA-N |
| PubChem CID | 233147 |
| IUPAC Name | 5-hydroxy-2,3-dihydroinden-1-one |
| SMILES | C1CC(=O)C2=C1C=C(C=C2)O |
Chemical Identifiers
| 3470-49-3 | |
| 148.161 | |
| ZRKQOVXGDIZYDS-UHFFFAOYSA-N | |
| 5-hydroxy-2,3-dihydroinden-1-one |
| C9H8O2 | |
| MFCD00857527 | |
| 233147 | |
| C1CC(=O)C2=C1C=C(C=C2)O |
Specifications
| 3470-49-3 | |
| C9H8O2 | |
| 5 g | |
| C1CC(=O)C2=C1C=C(C=C2)O | |
| 148.161 | |
| 148.16 | |
| Crystalline Powder |
| 184°C | |
| MFCD00857527 | |
| ZRKQOVXGDIZYDS-UHFFFAOYSA-N | |
| 5-hydroxy-2,3-dihydroinden-1-one | |
| 233147 | |
| ≥98.0% (GC,T) | |
| 5-Hydroxy-1-indanone |
Safety and Handling
TSCA : No