CAS RN 3470-49-3
IUPAC Name:
5-hydroxy-2,3-dihydro-1H-inden-1-one
Synonyms:
5-hydroxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol):
148.16
Molecular Formula:
C9H8O2
InChi Key:
ZRKQOVXGDIZYDS-UHFFFAOYSA-N
SMILES:
OC1=CC=C2C(=O)CCC2=C1
1
–
1
of
1
results
1
5-Hydroxy-1-indanone 98.0+%, TCI America™
CAS: 3470-49-3 Molecular Formula: C9H8O2 Molecular Weight (g/mol): 148.161 MDL Number: MFCD00857527 InChI Key: ZRKQOVXGDIZYDS-UHFFFAOYSA-N PubChem CID: 233147 IUPAC Name: 5-hydroxy-2,3-dihydroinden-1-one SMILES: C1CC(=O)C2=C1C=C(C=C2)O