missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxy-1-indanone 98.0+%, TCI America™
Supplier: TCI America H105125G
Specifications
| 5-Hydroxy-1-indanone | |
| 184°C | |
| MFCD00857527 | |
| ZRKQOVXGDIZYDS-UHFFFAOYSA-N | |
| 5-hydroxy-2,3-dihydroinden-1-one | |
| 233147 | |
| ≥98.0% (GC,T) |
| 3470-49-3 | |
| C9H8O2 | |
| 25 g | |
| C1CC(=O)C2=C1C=C(C=C2)O | |
| 148.161 | |
| 148.16 | |
| Crystalline Powder |
Chemical Identifiers
| 3470-49-3 | |
| 148.161 | |
| ZRKQOVXGDIZYDS-UHFFFAOYSA-N | |
| 5-hydroxy-2,3-dihydroinden-1-one |
| C9H8O2 | |
| MFCD00857527 | |
| 233147 | |
| C1CC(=O)C2=C1C=C(C=C2)O |
Safety and Handling
TSCA : No