missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenoic Acid 98.0+%, TCI America™

Supplier:  TCI America H087525G

Quantity 25 g

 View more versions of this product

Catalog No. H087525G


Only null left

Description

Description

Specifications

Chemical Identifiers

1577-22-6
114.144
XUDOZULIAWNMIU-UHFFFAOYSA-N
15308
C=CCCCC(=O)O
C6H10O2
MFCD00046558
5-hexenoic acid, delta-hexenoic acid, acmc-1bnq8
hex-5-enoic acid

Specifications

5-Hexenoic Acid
Colorless
C6H10O2
MFCD00046558
5-hexenoic acid, delta-hexenoic acid, acmc-1bnq8
C=CCCCC(=O)O
114.144
114.14
Liquid
1577-22-6
203°C
25 g
3265
XUDOZULIAWNMIU-UHFFFAOYSA-N
hex-5-enoic acid
15308
≥98.0% (GC,T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0609

TSCA : No

SDS
Documents

Documents

Promotions

Promotions