CAS RN 1577-22-6
IUPAC Name:
hex-5-enoic acid
Synonyms:
5-hexenoic acid delta-hexenoic acid
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
XUDOZULIAWNMIU-UHFFFAOYSA-N
SMILES:
OC(=O)CCCC=C
1
–
2
of
2
results
1
Promotions Available
5-Hexenoic acid, 99%
5-Hexenoic acid, 99%
CAS: 1577-22-6 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.144 MDL Number: MFCD00046558 InChI Key: XUDOZULIAWNMIU-UHFFFAOYSA-N Synonym: 5-hexenoic acid,delta-hexenoic acid,acmc-1bnq8 PubChem CID: 15308 IUPAC Name: hex-5-enoic acid SMILES: C=CCCCC(=O)O
2
5-Hexenoic Acid 98.0+%, TCI America™
CAS: 1577-22-6 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.144 MDL Number: MFCD00046558 InChI Key: XUDOZULIAWNMIU-UHFFFAOYSA-N Synonym: 5-hexenoic acid,delta-hexenoic acid,acmc-1bnq8 PubChem CID: 15308 IUPAC Name: hex-5-enoic acid SMILES: C=CCCCC(=O)O